Skip to Content
Merck
All Photos(3)

Key Documents

MABE977

Sigma-Aldrich

Anti-E3 ubiquitin-protein ligase UHRF1, clone BF18.11 Antibody

clone BF18.11, from mouse

Synonym(s):

E3 ubiquitin-protein ligase UHRF1, Nuclear protein 95, Nuclear zinc finger protein Np95, Ubiquitin-like PHD and RING finger domain-containing protein 1, mUhrf1, Ubiquitin-like-containing PHD and RING finger domains protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€316.00

€316.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€316.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€316.00


Estimated to ship on30 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BF18.11, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Uhrf1(18140)

General description

Ubiquitin-like with PHD and ring finger domains 1 (UHRF1), also known as ICBP90 or Np95, is a multifunctional nuclear protein that may be an important mediator of heterochromatin formation which is indicative of gene repression. UHRF1 contains a set and RING-finger-associated domain that detects hemi-methylated cytosine residues leading to the recruitment of DNMT1 to these residues during the S phase. UHRF1 thereby facilitates the transfer of methylation status to newly synthesized DNA molecules. UHRF1 may also interact with histones and HDAC1 via its PHD domain to promote modifications of histone residues which contribute to heterochromatin formation. UHRF1 may also directly modify histone H3 by ubiquitination. In addition, UHRF1 may contribute to the DNA-repair process though associations with PCNA. Several studies have suggested that overexpression of UHRF1 may contribute to the development of many cancers.

Immunogen

His-tagged recombinant protein corresponding to mouse E3 ubiquitin-protein ligase UHRF1.

Application

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected E3 ubiquitin-protein ligase UHRF1 in NIH/3T3 cells.
Immunoprecipitation Analysis: A representative lot immunoprecipitated E3 ubiquitin-protein ligase UHRF1 in 5 mg of NIH/3T3 cell lysate (Roberto Papait, FIRC Institute of Molecolar Oncology).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-E3 ubiquitin-protein ligase UHRF1, clone BF18.11 Antibody is validated for use in Western Blotting and Immunocytochemistry and Immunoprecipitation for the detection of E3 ubiquitin-protein ligase UHRF1.

Quality

Evaluated by Western Blotting in NIH-3T3 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected E3 ubiquitin-protein ligase UHRF1 in 10 µg of NIH-3T3 cell lysate.

Target description

~88 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service