Skip to Content
Merck
All Photos(1)

Key Documents

MABE1877

Sigma-Aldrich

Anti-EBNA-1 Antibody, clone 1EB14

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€146.00
100 μG
€369.00

€146.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€146.00
100 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

€146.00


Estimated to ship on27 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1EB14, monoclonal

mol wt

calculated mol wt 56.43 kDa
observed mol wt ~60 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal half

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

vaccinia virus ... EBNA-1(3783774)

General description

Epstein-Barr nuclear antigen 1 (UniProt: P03211; also known as EBNA-1, EBV nuclear antigen 1) is encoded by the EBNA1 (also known as BKRF1) gene (Gene ID: 3783709) in Epstein-Barr virus (EBV). EBV is a gamma-herpesvirus, a causative agent of infectious mononucleosis. EBNA1, a multi-functional protein, plays an important role in maintaining EBV in the host cell and has been implicated in host cell immortalization. It plays an essential role in replication and partitioning of viral genomic DNA during latent viral infection. During this phase, the circular double-stranded viral DNA undergoes replication once per cell cycle and is efficiently partitioned to the daughter cells. EBNA1 activates the initiation of viral DNA replication through binding to specific sites in the viral latent origin of replication, oriP. It is also reported to direct the segregation of viral episomes by mediating their attachment to host cell metaphase chromosomes and is known to activate the transcription of several viral latency genes. By counteracting the stabilization of host p53, it also reduces apoptosis and increases host cell survival. EBNA1 is also a risk factor in a variety of malignancies and is the only viral protein found in all EBV-related malignancies. Its C-terminal region contains a dimerization domain and a DNA binding domain (aa 459-607). Although this domain is essential for its replication function of EBNA1, it is not sufficient for this activity and the N-terminal half of EBNA1 is also required. (Ref.: Sun, X., et al. (2010). Proc. Natl. Acad. Sci. USA. 107(7); 3146-3151; Duellman, SJ., and Burgess, RR. (2009). PLoS ONE 4(2): e4614; Wu, H., et al. (2002). J. Virol. 76(5); 2480-2490).

Specificity

Clone 1EB14 is a mouse monoclonal antibody that detects Epstein-Barr nuclear antigen 1 (EBNA-1). It targets an epitope within 13 amino acids from the C-terminal half.

Immunogen

His-tagged recombinant Epstein-Barr nuclear antigen 1 expressed in E. coli.

Application

Quality Control Testing

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected EBNA-1 in Raji cell lysate.

Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated EBNA-1 in Immunoprecipitation application. (Duellman, S.J., et al. (2009). PLoS One. 4(2):e4614).

ELISA Analysis: A representative lot detected EBNA-1 in ELISA application. (Duellman, S.J., et al. (2009). PLoS One. 4(2):e4614).

Western Blotting Analysis: A representative lot detected EBNA-1 in Western Blotting application. (Duellman, S.J., et al. (2009). PLoS One. 4(2):e4614; Duellman, S. J., et al. (2006). Protein Expr Purif. 47(2):434--40).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-EBNA-1, clone 1EB14, Cat. No. MABE1877, is a mouse monoclonal antibody that detects Epstein-Barr nuclear antigen 1 and is tested in ELISA, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service