Skip to Content
Merck
All Photos(1)

Documents

MAB10219

Sigma-Aldrich

Anti-Epstein Barr Virus Antibody, clone 72A1 | MAB10219

ascites fluid, clone 72A1, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

72A1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG1

shipped in

dry ice

General description

Epstein-Barr virus, frequently referred to as EBV, is a member of the herpesvirus family and one of the most common human viruses. Although the symptoms of infectious mononucleosis usually resolve in 1 or 2 months, EBV remains dormant or latent in a few cells in the throat and blood for the rest of the person′s life. Periodically, the virus can reactivate and is commonly found in the saliva of infected persons. This reactivation usually occurs without symptoms of illness. EBV also establishes a lifelong dormant infection in some cells of the body′s immune system.

Specificity

The antibody reacts with the 350kDa/220kDa dalton viral envelope glycoprotein.

Immunogen

Disrupted fixed Epstein-Barr virus particles

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites
Liquid ascites fluid containing no preservatives.

Storage and Stability

1 year at -20°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monoclonal antibody against a 250,000-dalton glycoprotein of Epstein-Barr virus identifies a membrane antigen and a neutralizing antigen
Hoffman, G J, et al
Proceedings of the National Academy of Sciences of the USA, 77, 2979-2983 (1980)
Keiko Nagata et al.
Viral immunology, 31(8), 540-547 (2018-09-18)
Immunoglobulin (Ig) G4-related disease (IgG4-RD) is a newly recognized systemic fibroinflammatory disease with characteristic histological findings and high serum IgG4 levels. Epstein-Barr virus (EBV) is a persistent herpesvirus in B lymphocytes, and we previously reported EBV reactivation-induced Ig production. We
María López-Montañés et al.
Journal of immunology (Baltimore, Md. : 1950), 199(2), 656-665 (2017-06-21)
NK cells have been reported to respond against EBV-infected B cells in the lytic cycle and to control the viral infection involving IFN-γ secretion. Early reports proposed a role for NK cell Ab-dependent cellular cytotoxicity (ADCC) triggered via FcγR-IIIA (CD16)
Chien-Hui Hung et al.
Journal of virology, 88(20), 12133-12145 (2014-08-15)
Autophagy is an intracellular degradation pathway that provides a host defense mechanism against intracellular pathogens. However, many viruses exploit this mechanism to promote their replication. This study shows that lytic induction of Epstein-Barr virus (EBV) increases the membrane-bound form of
Keiko Nagata et al.
Autoimmunity, 48(5), 328-335 (2015-03-12)
Epstein-Barr virus (EBV) is a ubiquitous virus that infects most adults latently. It persists in B lymphocytes and reactivates occasionally. Graves' disease is an autoimmune hyperthyroidism caused by thyrotropin receptor antibodies (TRAbs). We have reported that Graves' disease patients and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service