Skip to Content
Merck
All Photos(2)

Documents

MABD106

Sigma-Aldrich

Anti-OTX2 Antibody, clone 8D8.2

clone 8D8.2, from mouse

Synonym(s):

Homeobox protein OTX2, Orthodenticle homolog 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.75

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8D8.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... OTX2(5015)

General description

Homeobox protein OTX2 (UniProt P32243; also known as Orthodenticle homolog 2) is encoded by the OTX2 (also known as CPHD6, MCOPS5) gene (Gene ID 5015) in human. OTX2 is a paired-type homeobox transcription factor that controls the expression of target genes by binding to regulatory regions containing the YTAATNN motifs. Studies conducted in mice show developmental stage-dependent OTX2 expression pattern. OTX2 is widely expressed in the epiblast, but becomes progressively regionalized during gastrulation to an anterior domain of the ectoderm (anterior neuroectoderm or ANE) that contains the precursor cells of the prosencephalon and the mesencephalon of the embryonic brain. The precursor cells of the anterior visceral endoderm (AVE) fail to translocate from the distal site to the anterior region of the pre-gastrulation stage Otx2-knockout embryos. The absence of the AVE, in turn, causes the loss of the source of morphogenetic signals for inducing neural differentiation of the ANE. In addition, OTX2 is shown to be essential in the anterior mesendoderm (AME) for the induction of anterior neural tissues and head formation. OTX2 is reported to mediate the transcriptions of Dkk1 and Lhx1, loss of Otx2 in the AME results in the down regulation of Dkk1 and Lhx1.

Specificity

Expected to react with both spliced isoforms of human OTX2 reported by UniProt (P32243).

Immunogen

Epitope: Internal.
GST-tagged recombinant human OTX2 internal fragment.

Application

Detect OTX2 using this Anti-OTX2 Antibody, clone 8D8.2 validated for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected OTX2 in human placenta and retina tissue sections.
Research Category
Stem Cell Research
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in human retinoblastoma Y79 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected OTX2 in 10 µg of human retinoblastoma Y79 cell lysate.

Target description

~32 kDa observed. 31.64 kDa (isoform 1) and 32.41 kDa (isoform 2) calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service