Skip to Content
Merck
All Photos(1)

Key Documents

MABC82

Sigma-Aldrich

Anti-RIP1/RIPK1 Antibody, clone 3C6.1

clone 3C6.1, from mouse

Synonym(s):

Receptor-interacting serine/threonine-protein kinase 1, Cell death protein RIP, Receptor-interacting protein 1, RIP-1, Serine/threonine-protein kinase RIP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3C6.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RIPK1(8737)

General description

Receptor-interacting serine/threonine-protein kinase 1, also known as Cell death protein RIP or Receptor-interacting protein 1 (RIP-1) or Serine/threonine-protein kinase RIP, and encoded by the gene name RIPK1 or RIP or RIP1, contains 1 death domain and 1 protein kinase domain. RIPK1 is localized to the cytoplasm and plays an important role in a variety of cellular pathways including the NF-κB pathway and programmed necrotic cell death (necroptosis).

Immunogen

GST-tagged recombinant protein corresponding to human RIP1/RIPK1.

Application

Detect RIP1/RIPK1 using this Anti-RIP1/RIPK1 Antibody, clone 3C6.1 validated for use in western blotting.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RIP1/RIPK1 in 200 µg of Jurkat cell lysate.

Target description

~ 76 kDa observed

Linkage

Replaces: MABC688

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service