Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1767

Sigma-Aldrich

Anti-RIP1/RIPK1 Antibody, clone 1N16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1N16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 75.93 kDa
observed mol wt ~40 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RIPK1(8737)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1N16 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Receptor-interacting serine/threonine-protein kinase 1 (RIP1). It targets an epitope within 22 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the N-terminal half of human Receptor-interacting serine/threonine-protein kinase 1 (RIP1).

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RIP1/RIPK1 in NIH3T3 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected RIP1/RIPK1 in MCF7, A549, and Raw264.7 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected RIP1/RIPK1 in human small intestine tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound RIP/RIPK1 peptide with a KD of 2.3 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RIP1/RIPK1 in HT-29 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Receptor-interacting serine/threonine-protein kinase 1 (UniProt: Q13546; also known as EC:2.7.11.1, Cell death protein RIP, Receptor-interacting protein 1, RIP-1, RIPK1) is encoded by the RIPK1 (also known as RIP, RIP1) gene (Gene ID: 8737) in human. RIPK1 is a serine-threonine kinase that serves as a key regulator of TNF-a-mediated apoptosis, necroptosis, and inflammatory pathways. It exhibits kinase activity-dependent functions that regulates cell death and kinase-independent scaffold functions that regulate inflammatory signal transduction and cell survival. Upon binding of TNF-a to TNFR1, RIPK1 is recruited to the TNF-R1 signal transduction complex where it acts as a scaffold protein promoting cell survival, in part, by activating the canonical NF-kB pathway. RIPK1 is required to limit caspase 8-dependent TNFR1-induced apoptosis. Under normal conditions, RIPK1 acts as an inhibitor of RIPK3-dependent necroptosis via FADD-mediated recruitment of caspase 8, which cleaves RIPK1 (at Asp 324) and limits TNF-a-induced necroptosis. It is also required to inhibit apoptosis and necroptosis during embryonic development where it acts by preventing the interaction of TRADD with FADD thereby limiting aberrant activation of caspase 8. In addition to apoptosis and necroptosis, it is also involved in inflammatory response by promoting transcriptional production of pro-inflammatory cytokines. RIPK1 and RIPK3 are known to undergo reciprocal auto- and trans-phosphorylation. Phosphorylation of RIPK1 at Ser 161 by RIPK3 is crucial for the formation of the necroptosis-inducing complex. Its phosphorylation at Ser 25 represses its kinase activity and consequently prevents TNF-a-mediated RIPK1-dependent cell death. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Dondelinger, Y., et al. (2019). Nat. Commun. 10(1); 1729; Meng, H., et al. (2018). Proc. Natl. Acad. Sci. USA. 115(9); E2001-E2009; He, S., et al. (2009). Cell. 137(6); 1100-1111).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service