Skip to Content
Merck
All Photos(3)

Documents

MABC609

Sigma-Aldrich

Anti-Thymidylate Kinase/TMPK Antibody, clone 1A5.1

clone 1A5.1, from mouse

Synonym(s):

Thymidylate kinase, Thymidylate Kinase/TMPK, dTMP kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A5.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TK1(7083)

General description

Thymidylate kinase (EC 2.7.4.9; UniProt P23919; also known as dTMP kinase) is encoded by the DTYMK (also known as CDC8, PP3731, TMPK, TYMK) gene (Gene ID 1841) in human. Thymidylate kinase (TMPK) is an ubiquitously expressed enzyme of the dTTP synthesis pathway involved in DNA synthesis. TMPK catalyzes the phosphorylation of thymidine 5′-monophosphate (dTMP) in the presence of ATP and Mg(2+) to form thymidine 5′-diphosphate (dTDP), which is then further phosphorylated by nucleoside-diphosphate kinase (NDK) to thymidine 5′-triphosphate (dTTP). Aside from its normal physiological role, TMPK activity is essential for the activation of nucleoside analogs-based prodrugs, such as the clinically used anti-HIV prodrugs AZT and d4T, which need to be converted to the triphosphate forms for their pharmacological activities. Inhibition of TMPK activity is reported to augment the potency of anticancer agent doxorubicin against human colon cancer cells regardless of their p53 status. In addition, lung cancers with LKB1-mutations are reported to show deficits in nucleotide metabolism and exhibit hypersensitivity to DTYMK inhibition.

Immunogen

GST-tagged recombinant protein corresponding to human Thymidylate Kinase/TMPK.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-TMPK Antibody, clone 1A5.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of Thymidylate Kinase .
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Thymidylate Kinase/TMPK in 10 µg of MCF-7 cell lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Thymidylate Kinase/TMPK in human breast cancer and human ovarian cancer tissue.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Thymidylate Kinase/TMPK in 10 µg of Jurkat cell lysate.

Target description

~24 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service