Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Check Cart for Availability | € 416,00 |
About This Item
UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IHC, IP, WB
Citations:
-
conjugate
unconjugated
Quality Segment
antibody form
purified antibody
clone
polyclonal
species reactivity
invertebrates
technique(s)
ELISA: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
UniProt accession no.
target post-translational modification
unmodified
General description
Luciferase [Photinus pyralis (Firefly)].
Application
Epitope: Firefly
Biochem/physiol Actions
Luciferase [Photinus pyralis (Firefly)]. Monospecific by IEP. May show cross reactivity with Luciferase from other sources.
Physical form
Immunoglobulin purified by delipidation, salt fractionation and ion exchange chromatography. Liquid in 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% sodium azide.
Preparation Note
Maintain frozen at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.