Skip to Content
Merck

Skip To

MABC538

Anti-FANCJ/BACH1 Antibody, clone 2G7

clone 2G7, from mouse

Synonym(s):

Fanconi anemia group J protein, Protein FACJ, ATP-dependent RNA helicase BRIP1, BRCA1-associated C-terminal helicase 1, BRCA1-interacting protein C-terminal helicase 1, BRCA1-interacting protein 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 463,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
2G7, monoclonal
Application:
ICC, IP, WB
Citations:
1

€ 463,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G7, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BRIP1(83990)

General description

Bach1 and Bach2 constitute a subfamily of the basic region-leucine zipper family. Bach proteins form heterodimers with small Maf proteins, bind to the Maf recognition element (MARE) and repress target gene expression. Transcription regulator protein BACH2 mediates early B cell differentiation and is specifically expressed in pro and mature B-cells. This protein plays an important role in both the immunoglobulin class switch, oncogene-induced senescence, and somatic hypermutation.
~140 kDa observed

Immunogen

Epitope: C-terminus
Linear peptide corresponding to the C-terminus of human FANCJ/BACH1.

Application

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected FANCJ/BACH1 in MCF-7 cells (Cantor, S. B., et al. (2001). Cell. 105(1):149-160.).

Western Blotting Analysis: A representative lot from an independent laboratory detected FANCJ/BACH1 in transfected FA-J cell lysate (Xie, J., et al. (2010). Oncogene. 29(17):2499-2508.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated FANCJ/BACH1 from MCF-7 cell lysate, prior to detecting the immunoprecipitated sample in Western Blot (Cantor, S. B., et al. (2001). Cell. 105(1):149-160.).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-FANCJ/BACH1 Antibody, clone 2G7 is validated for use in western blotting, ICC & IP for the detection of FANCJ/BACH1.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected FANCJ/BACH1 in 10 µg of MCF-7 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC524MABE1069SAB5700340
species reactivity

human

species reactivity

mouse, human, rat

species reactivity

human

species reactivity

rat, human, mouse

clone

2G7, monoclonal

clone

8F3, monoclonal

clone

3C11, monoclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit

Gene Information

human ... BRIP1(83990)

Gene Information

human ... FANCC(2176)

Gene Information

human ... BAP1(8314)

Gene Information

human ... BRCA1(672)

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters