Skip to Content
Merck
All Photos(2)

Key Documents

MABC524

Sigma-Aldrich

Anti-FANCC Antibody, clone 8F3

clone 8F3, 1 mg/mL, from mouse

Synonym(s):

Fanconi anemia group C protein, Protein FACC, FANCC

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€435.00

€435.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€435.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€435.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8F3, monoclonal

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FANCC(2176)

General description

Fanconi anemia (FA) is an inherited bone marrow failure syndrome associated with a progressive decline in hematopoietic stem cells, developmental defects, and predisposition to cancer. FANCC is part of the multisubunit Fanconi anemia (FA) complex composed of FANCA, FANCB, FANCC, FANCE, FANCF, FANCG, FANCL/PHF9 and FANCM. FANCC is a primarily nuclear localized DNA repair protein that is believed to function in interstrand DNA cross-link repair and the maintenance of normal chromosome stability. Mutations in FANCC are associated with various aspects of FA. New research shows that FANCC is critical for resistance to DNA cross-linking drug-induced DNA damage in cells patients with myeloproliferative neoplasms. Also, FANCC interacts with the C-terminal-binding protein-1 (CtBP1) involved in transcriptional regulation, including being a negative regulator of Dickkopf-1 (DKK1) expression. This down regulation does not occur in mutations of FANCC, which then leads to up-regulation of DKK1, a molecule involved in hematopoietic malignancies.

Immunogen

Linear peptide corresponding to Human FANCC.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-FANCC Antibody, clone 8F3 is validated for use in Western Blotting and Immunoprecipitation and Immunocytochemistry for the detection of FANCC.
Western Blotting Analysis: 4 µg/mL of a representative lot of this antibody detected FANCC in 10 µg of Mouse brain lysate.

Western Blotting Analysis: A representative lot detected FANCC in HeLa cell lysate (Tremblay, C.S., et al. (2008) Blood. 112(5):2062-2070).

Immunoprecipitation Analysis: A representative lot immunoprecipitated FANCC from HeLa cell lysate (Tremblay, C.S., et al. (2008) Blood. 112(5):2062-2070).

Immunocytochemistry Analysis: A representative lot detected FANCC in HeLa cells (Tremblay, C.S., et al. (2008) Blood. 112(5):2062-2070).

Quality

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected FANCC in 10 µg of PC12 cell lysate.

Target description

~ 63 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FengFei Huang et al.
BMC research notes, 11(1), 662-662 (2018-09-15)
The Uncoordinated 5A (UNC5A) protein is part of a family of receptors that play roles in axonal pathfinding and cell migration. We previously showed that the Fanconi anemia C protein (FANCC) interacts with UNC5A and delays UNC5A-mediated apoptosis. FANCC is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service