Skip to Content
Merck
All Photos(1)

Key Documents

MAB8537

Sigma-Aldrich

Anti-Herpes Virus 6 Antibody, A & B, gp 60/110

Chemicon®, from mouse

Synonym(s):

HHV6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€424.00

€424.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€424.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€424.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

shipped in

wet ice

Specificity

Specific for the gp 60/110 envelope glycoprotein of HHV6.

CELLULAR LOCALIZATION:

Cytoplasmic and membrane

STRAIN RECOGNITION:

Types A & B

Immunogen

Epitope: A & B, gp 60/110

Application

Detect Herpes Virus 6 using this Anti-Herpes Virus 6 Antibody, A & B, gp 60/110 validated for use in IF, IP & WB.
Western Blot 1:10 - 1:50

Immunofluorescence 1:10-1:50

Immunoprecipitation 1:10-1:50

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.01 M phosphate buffered saline (PBS) pH 7.2, containing 0.1% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence for human herpesvirus 6 variant A antibodies in multiple sclerosis: diagnostic and therapeutic implications.
J O Virtanen, M Farkkila, J Multanen, L Uotila, A J Jaaskelainen, A Vaheri, M Koskiniemi
Journal of Neurovirology null
Purification of infectious human herpesvirus 6A virions and association of host cell proteins.
Hammarstedt, M; Ahlqvist, J; Jacobson, S; Garoff, H; Fogdell-Hahn, A
Virology Journal null
Kageaki Kuribayashi et al.
Internal medicine (Tokyo, Japan), 45(7), 475-478 (2006-05-09)
A 31-year-old man referred to our hospital for treatment of his chronic myeloid leukemia (CML) in the first chronic phase by bone marrow transplantation. We pretreated him with cyclophosphamide and total body irradiation and bone marrow transplantation (BMT) was carried

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service