Skip to Content
Merck
All Photos(2)

Key Documents

SAB2108453

Sigma-Aldrich

Anti-TFEB

affinity isolated antibody

Synonym(s):

Anti-Math5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€524.00

€524.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€524.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€524.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

horse, human, rabbit, pig, mouse, goat, bovine, rat

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFEB(7942)

General description

Transcription factor EB (TFEB) is encoded by the gene mapped to human chromosome 6p21.1. The encoded protein belongs to the MiT transcription factor family. TFEB is widely expressed and is characterized with a transactivation domain, basic-helix-loop-helix domain, glutamine rich region, leucine zipper region and proline rich region.

Immunogen

Synthetic peptide directed towards the middle region of human TFEB

Biochem/physiol Actions

Transcription factor EB (TFEB) plays a key role in the regulation of autophagy and lysosome biogenesis. TFEB, expressed in endothelial cells, reduces inflammation and hinders atherosclerosis development. Thus, this protein can be considered as a potent therapeutic target for atherosclerosis and associated cardiovascular diseases.

Sequence

Synthetic peptide located within the following region: IKELGMLIPKANDLDVRWNKGTILKASVDYIRRMQKDLQKSRELENHSRR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Transcription Factor EB Links Cellular Stress to the Immune Response
Nabar NR, et al.
The Yale Journal of Biology and Medicine, 90(2), 301-315 (2017)
Molecular Genetics and Cellular Characteristics of TFE3 and TFEB Translocation Renal Cell Carcinomas
Kauffman EC, et al.
Nature reviews. Urology, 11(8), 465-465 (2014)
TFEB inhibits endothelial cell inflammation and reduces atherosclerosis.
Lu H, et al.
Science Signaling, 10(464) (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service