Skip to Content
Merck
All Photos(2)

Documents

MAB8152-I

Sigma-Aldrich

Anti-West Nile Virus/Kunjin Antibody, NS1 Antibody, clone 3.1112G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3.1112G, monoclonal

mol wt

calculated mol wt 381.37 kDa

species reactivity

virus

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgM

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

vaccinia virus ... poly> POLY(912267)

General description

Genome polyprotein (UniProt: P14335) is encoded by the POLY gene (Gene ID: 912267) in West Nile virus. West Nile virus (WNV) is a mosquito-borne positive-sense RNA virus of the Flavivirus genus within the Flaviviridae family. WNV genome encodes for three structural domains known as C, prM, and E and seven non-structural proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B, and NS5). These are translated as a single polypeptide, which is subsequently cleaved by viral and cellular proteases. NS1 (aa 792-1143) is a glycoprotein that is involved in immune evasion, pathogenesis, and viral replication. It is secreted from infected cells and may vary its oligomeric state between monomers, dimers (the primary form) and hexamers. Once cleaved off the polyprotein, it is targeted to three destinations: the viral replication cycle, the plasma membrane, and the extracellular compartment. Intracellular NS1 functions as an essential cofactor for viral replication, and it localizes to WNV replication sites, whilst cell surface and secreted NS1 act as immunomodulators. NS1 is required for formation of the replication complex and recruitment of other non-structural proteins to the ER-derived membrane structures. It is reported to antagonize the complement function and inhibit signal transduction originating from Toll-like receptor 3. (Ref.: Martín-Acebes MA, et al. (2012). World J Virol. 1(2); 51-70).

Specificity

Clone 3.1112G is a mouse monoclonal antibody that detects West Nile virus NS1 protein.

Immunogen

West Nile Virus (WNV; subtype KUNV) (isolate OR393).

Application

Quality Control Testing

Evaluated by ELISA with recombinant West Nile Virus NS1 protein.

ELISA Analysis: Various concentrations of this antibody detected recombinant West Nile virus NS1 protein.

Tested Applications

ELISA Analysis: A representative lot detected West Nile Virus/Kunjin, NS1 in ELISA applications (Shelite, T.R., et al. (2008). Vector Borne Zoonotic Dis.;8(3):321-9; Ledermann, J.P., et al. (2011). Clin Vaccine Immunol.;18(4):580-7; Thompson, N.N., et al. (2012). Vector Borne Zoonotic Dis.;12(11):969-78).

Affinity Binding Assay: A representative lot of this antibody bound West Nile Virus/Kunjin with a KD of 1.0 x 10-12 in an affinity binding assay.

Western Blotting Analysis: A representative lot detected West Nile Virus/Kunjin, NS1 in Western Blotting applications (Whiteman, M.C., et al. (2011). Vaccine.;29(52):9702-10).

Immunofluorescence Analysis: A representative lot detected West Nile Virus/Kunjin, NS1 in Immunofluorescence applications (Garcia-Tapa, D. et al. (2006). Vet Immunol Immunopathol.;110(3-4):229-44; Giel-Moloney, M., et al. (2019). Sci Rep.;9(1):20005).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-West Nile Virus/Kunjin, NS1, clone 3.1112G, Cat. No. MAB8152-I, is a mouse monoclonal antibody that detects NS1 protein of West Nile virus and is tested for use in Affinity Binding Assay, ELISA, Immunofluorescence, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service