Merck
  • Antibodies and methods for immunohistochemistry of extracellular matrix proteins.
Antibodies and methods for immunohistochemistry of extracellular matrix proteins.
Matrix biology : journal of the International Society for Matrix Biology (2018-05-08)
Steffen Rickelt, Richard O Hynes
ABSTRACT
The diversity of extracellular matrix (ECM) proteins encoded in mammalian genomes and detected by proteomic analyses generates a need for well validated antibodies against these proteins. We present characterization of a large number of antibodies against ECM proteins, from both commercial and academic sources, together with discussion of methods and strategies for their effective use in immunohistochemistry and illustrations of their efficacy. These data should be of value to investigators seeking well validated antibodies to ECM proteins of interest and save significant time and money tracking down effective reagents.
Sigma-Aldrich
Anti-EMILIN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Collagen Type VI Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Versican Antibody, a.a. 1360-1439 of mouse versican, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Agrin Antibody, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glypican-1 Antibody, clone 4D1, clone 4D1, from mouse
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-TGFBI antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Fibronectin, Cellular antibody produced in mouse, clone FN-3E2, ascites fluid
Sigma-Aldrich
Anti-Heparan Sulfate Proteoglycan (Perlecan) Antibody, clone 5D7-2E4, clone 5D7-2E4, from mouse
Sigma-Aldrich
Anti-Mouse Collagen Type I Antibody, Chemicon®, from rabbit