Skip to Content
Merck
All Photos(1)

Key Documents

MAB2502

Sigma-Aldrich

Anti-Fibrillin-1 Antibody, NT, clone 26

clone 26, Chemicon®, from mouse

Synonym(s):

Anti-ACMICD, Anti-FBN, Anti-GPHYSD2, Anti-MASS, Anti-MFLS, Anti-MFS1, Anti-OCTD, Anti-SGS, Anti-SSKS, Anti-WMS, Anti-WMS2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€557.00

€557.00


Estimated to ship on28 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1019

Request a Bulk Order

Select a Size

Change View
100 μG
€557.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€557.00


Estimated to ship on28 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1019

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

26, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FBN1(2200)

Specificity

Monoclonal antibody MAB2502 recognizes human Fibrillin-1. Epitope mapping studies identify the binding site of this antibody to amino-terminal end of the molecule, between amino acid residues 45 and 450. The antibody is reactive with human, chicken, and bovine Fibrillin-1.

Immunogen

Epitope: N-terminus
Human Fibrillin-1

Application

Anti-Fibrillin-1 Antibody, N-terminus, clone 26 detects level of Fibrillin-1 & has been published & validated for use in ELISA, IP, WB & IC.
Immunoblotting

Immunofluorescence

Immunoprecipitation

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Liquid at 1 mg/mL in 20 mM phosphate buffer, 250 mM NaCl, pH 7.6, containing 0.1% sodium azide.Note: Sodium azide is toxic. MSDS available upon request.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James C Tan et al.
Scientific reports, 14(1), 3517-3517 (2024-02-13)
Aqueous humor (AH) and blood levels of transforming growth factor β (TGFβ) are elevated in idiopathic primary open angle glaucoma (POAG) representing a disease biomarker of unclear status and function. Tsk mice display a POAG phenotype and harbor a mutation
Natalija Bogunovic et al.
Journal of endovascular therapy : an official journal of the International Society of Endovascular Specialists, 28(4), 604-613 (2021-04-28)
Abdominal aortic aneurysms (AAAs) are associated with overall high mortality in case of rupture. Since the pathophysiology is unclear, no adequate pharmacological therapy exists. Smooth muscle cells (SMCs) dysfunction and extracellular matrix (ECM) degradation have been proposed as underlying causes.
Nagako Yoshiba et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 63(6), 438-448 (2015-03-26)
Myofibroblasts and extracellular matrix are important components in wound healing. Alpha-smooth muscle actin (α-SMA) is a marker of myofibroblasts. Fibrillin-1 is a major constituent of microfibrils and an extracellular-regulator of TGF-β1, an important cytokine in the transdifferentiation of resident fibroblasts
Fibrillin microfibril structure identifies long-range effects of inherited pathogenic mutations affecting a key regulatory latent TGFI?-binding site.
Godwin, et al.
Nature Structural and Molecular Biology, 30, 608-618 (2023)
Rachel Morissette et al.
The Journal of clinical endocrinology and metabolism, 100(8), E1143-E1152 (2015-06-16)
The contiguous gene deletion syndrome (CAH-X) was described in a subset (7%) of congenital adrenal hyperplasia (CAH) patients with a TNXA/TNXB chimera, resulting in deletions of CYP21A2, encoding 21-hydroxylase necessary for cortisol biosynthesis, and TNXB, encoding the extracellular matrix glycoprotein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service