Skip to Content
Merck
All Photos(1)

Documents

SAB4200718

Sigma-Aldrich

Anti-Elastin antibody, Mouse monoclonal

clone BA-4, purified from hybridoma cell culture

Synonym(s):

Anti-ELN, Anti-Tropoelastin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

BA-4, monoclonal

form

buffered aqueous solution

species reactivity

sheep, hamster, bovine, goat, frog, rabbit, porcine, human, guinea pig, canine

concentration

~1.0 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: 10-20 μg/mL using enzyme-retrieved formalin-fixed, paraffin-embedded sheep arterial tissue sections
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EMILIN3(2006)

General description

Anti-Elastin antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the BA-4 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with bovine alpha-Elastin.

Immunogen

bovine alpha-Elastin

Application

Anti-Elastin antibody, Mouse monoclonal has been used in:
  • immunohistochemistry
  • immunoblot
  • immunoprecipitation

Biochem/physiol Actions

Elastin (ELN) provides elasticity and is integrated with other proteins from the ECM in elastic fibrils. Elastin undergoes changes of morphology with aging and at many disease states. In addition, several heritable diseases, such as supravalvular aortic stenosis, cutis laxa, Marfan syndrome, pseudoxanthoma elasticum and Buschke-Ollendorf syndrome, may result from molecular defects of elastin.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elastin
Biochemistry of Collagens, Laminins and Elastin, 197-201 (2016)
Impaired elastogenesis in Hurler disease: dermatan sulfate accumulation linked to deficiency in elastin-binding protein and elastic fiber assembly
Hinek A and Wilson SE
The American Journal of Pathology, 156(3), 925-938 (2000)
Elastogenesis in human arterial disease: a role for macrophages in disordered elastin synthesis.
Krettek A, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 23(4), 582-587 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service