Skip to Content
Merck
All Photos(1)

Key Documents

MAB19294

Sigma-Aldrich

Anti-Integrin beta 1 (CD29) Antibody

CHEMICON®, mouse monoclonal, TASC/9D11

Synonym(s):

CD29

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€626.00

€626.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€626.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€626.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

Anti-Integrin β1 Antibody, active β1 integrins, clone TASC/9D11, clone TASC/9D11, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TASC/9D11, monoclonal

species reactivity

chicken

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

Specificity

Reactive with the extracellular domain of chicken integrin beta 1

Immunogen

E10 chick brain glial cells
Epitope: active beta1 integrins

Application

Detect the Integrin β1 protein using this Anti-Integrin β1, active β1 integrins, clone TASC/9D11 validated for use in IP & ICC.
Western blot: Not effective

Immunocytochemistry

Immunoprecipitation

Biological activity: Functionally inhibits attachment to vitronectin. Activates attachment to other substrates.



Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M PB +0.25 M NaCl, pH=7.6 with 0.1% Sodium Azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L R Strachan et al.
Experimental cell research, 314(3), 441-452 (2007-11-27)
Cell migration is essential for proper development of numerous structures derived from embryonic neural crest cells (NCCs). Although recent work has shown that receptor recycling plays an important role in NCC motility on laminin, the molecular mechanisms regulating NCC motility
Masayuki Yamashita
Communications biology, 6(1), 680-680 (2023-07-01)
Retinal ganglion cell (RGC) axons converge on the optic disc to form an optic nerve. However, the mechanism of RGC axon convergence remains elusive. In the embryonic retina, an electric field (EF) exists and this EF converges on the future
Ewa Mrówczyńska et al.
Frontiers in cell and developmental biology, 12, 1433947-1433947 (2024-08-15)
37/67 kDa laminin receptor (LamR)/ribosomal protein SA exhibits dual function as both a ribosomal protein and cell surface receptor for laminin. LamR influences critical cellular processes such as invasion, adhesion, and migration when acting as a receptor. Despite the acknowledged importance
Integrins and cAMP mediate netrin-induced growth cone collapse.
Lemons, ML; Abanto, ML; Dambrouskas, N; Clements, CC; Deloughery, Z; Garozzo, J; Condic, ML
Brain Research null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service