Skip to Content
Merck
All Photos(1)

Documents

IQ7XUVKT1

Milli-Q

ech2o® UV lamp

Protects stored water against biofilm buildup in Milli-Q® IQ storage tanks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
NACRES:
NB.33

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

ech2o®

compatibility

for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015

Looking for similar products? Visit Product Comparison Guide

General description

This ech2o® UV lamp kit includes 1 mercury-free UV LED bactericidal lamp for the Milli-Q® IQ 7003/05/10/15, 1 mercury-free UV LED bactericidal lamp for the ASM of Milli-Q® IQ/IX storage tanks and 1 mercury-free UV excimer lamp for oxidation of organic contaminants for Milli-Q® IQ 7003/05/10/15.

Application

The ech2o® UV lamp kit is used to limit the development of biofilm in system and storage tank, and to ensure oxidation of organic contaminants.

Features and Benefits

  • All three UV lamps are mercury-free
  • 2 lamps are with UVc LED technology (emitting at 265 nm) for inactivation of bacteria
  • 1 lamp is with excimer technology (emitting at 172 nm) for photo-oxidation of organic contaminants

Components

ech2o® lamp for the system & ech2o® ASM lamp for for the tank top & ech2o® lamp for oxidation of organic contaminants in the system

Legal Information

ECH2O is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service