Skip to Content
Merck
All Photos(8)

Key Documents

IQ700XPKIT

Milli-Q

Milli-Q® IQ 7003/05 Ultrapure Water Purification Kit

greener alternative

For Milli-Q® IQ 7003 and 7005 ultrapure water systems. This product replaces IQ700XPKT1.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
Pricing and availability is not currently available.

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Milli-Q®

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

humidity measuring range

10-80% (In original sealed packaging to protect purification elements)

H

39.5 cm (15.5 in.)

input

feed water nature potable tap water (within specifications of the water purification system)

compatibility

for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005

greener alternative category

shipped in

ambient

storage temp.

2-30°C

General description

This water purification kit includes:

  • IPAK Gard® 03/05 pretreatment cartridge composed of a pleated filter for the removal of particulates, minerals and colloids and a carbon block for the removal of free chlorine. This protects the reverse osmosis (RO) membrane and improves system performance.
  • IPAK Quanta® polishing cartridge for Elix® pure water contains innovative IQnano ion-exchange media for ion removal down to trace levels, plus high-grade synthetic activated carbon targets trace organic contaminants.
  • Tank vent filter to protect stored pure (Type 2) water from airborne contaminants. It contains activated carbon, soda lime and a membrane filter to prevent impure air from entering the tank.

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Application

  • For Milli-Q® IQ 7003/05 ultrapure water systems
  • This kit combines an IPAK Gard® pretreatment cartridge that protects RO membranes and IPAK Quanta® polishing cartridge for Elix® feed water that ensures the consistent production of ultrapure water
  • Tank vent filter protects stored pure (Type 2) water from airborne contaminants

Features and Benefits

  • e-Sure tags enable RFID connection with the system, for seamless consumable status monitoring, data management and full traceability
  • Supplied with one Drain Cap per cartridge, to remove excess water and reduce the weight of exhausted cartridges at disposal
  • 63% of the parts suppliers for the IPAK Gard® cartridge and 78% of the parts suppliers for IPAK Quanta® cartridge included in this kit are ISO 14001 environmental management system-certified

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service