Skip to Content
Merck
All Photos(4)

Documents

FR180380641

Sigma-Aldrich

Estapor® Carboxylated Colored Microspheres

K1-040 Blue Int COOH PS 5% Blue Colored Carboxylated microspheres 400nm, Lateral Flow Immunoassay

Synonym(s):

Colored Beads, Detector Beads, Dyes Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13102026
NACRES:
NA.24

material

polystyrene (microspheres)

Quality Level

packaging

bottle of 5—250 mL

manufacturer/tradename

Estapor®

concentration

5% solids

technique(s)

lateral flow assay: suitable (immunoassay)

color

bluevery strong

matrix

polymeric Poly-styrene-co-acrylate

bead diameter

0.340—0.440 μm

capacity

10—100 μeq/g surface charge density

λmax

654 nm

functional group

carboxylic acid (COOH)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Estapor® microspheres have been the top choice for many lateral flow diagnostic test manufacturers. Used in millions of assays made by internationally known brands, our microspheres are adapted to both both qualitative and quantitative Lateral Flow ImmunoAssays.

Application

Main applications of the Color Intense microspheres include qualitative and quantitive Laterflow ImmunoAssays (LFIA)

Features and Benefits

Key advantages of Estapor® Color Intense Microspheres include:

  • Improved assay sensitivity through greater signal intensity.
  • Easy reading, even at lower conjugate concentration.
  • Multiplexing ready by combining Red and Blue Intense allows an easy differentiation of test lines.

Legal Information

Estapor is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover qualitative tests with lateral flow assays, including multiplex tests. Learn about their fast, user-friendly applications in diagnostics.

Related Content

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service