FR180380638
Estapor® Carboxylated Colored Microspheres
K1-040 Red Int COOH PS 5%, Red Colored Carboxylated microspheres 400nm, Lateral Flow Immunoassay, LFIA, IVD - For ordering, click "Request more information"
Synonym(s):
Colored Beads, Detector Beads, Dyes Beads
About This Item
material
high-density polyethylene packaging (Bottle)
polystyrene beads
Quality Level
form
microspheres
packaging
bottle of 5-250 mL
manufacturer/tradename
Estapor®
concentration
5.0% solids
technique(s)
lateral flow assay: suitable (Immunoassay)
diam.
0.340-0.440 μm
color
redvery strong
matrix
polymeric Poly-styrene-co-acrylate
bead diameter
0.340-0.440 μm±<5%
capacity
10-100 μeq/g surface charge density
λmax
525 nm
functional group
carboxylic acid (COOH)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Improved assay sensitivity through greater signal intensity.
- Easy reading, even at lower conjugate concentration.
- Multiplexing ready by combining Red and Blue Intense allows an easy differentiation of test lines.
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover qualitative tests with lateral flow assays, including multiplex tests. Learn about their fast, user-friendly applications in diagnostics.
Related Content
Estapor Red Intense microspheres improve lateral flow test performance in HBsAg detection.
Discover how Estapor® Red Intense microspheres outperform colloidal gold in lateral flow immunoassays, enhancing sensitivity, speed, and reliability for diagnostic applications.
Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.
Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service