Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB207F

Sigma-Aldrich

Milli-Mark® Anti-CD71-FITC Antibody, clone Ber-T9

clone Ber-T9, Milli-Mark®, from mouse

Synonym(s):

Transferrin receptor protein 1, TfR1, TfR, T9, p90

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€370.00

€370.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€370.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ber-T9, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TFRC(7037)

General description

CD71 is a 190 kDa disulfide-bonded homodimeric type II transmembrane glycoprotein. CD71 is the receptor for transferrin. CD71 is involved in iron (Fe3+) uptake, and expression is regulated by the metabolic demand for iron, which is essential for the growing cell (Goding, 2002). CD71 is more abundantly expressed in rapidly dividing cells than quiescent cells (Newman, 1982; Sutherland, 1981). In normal tissues, high expression of CD71 is seen in the placental trophoblast and hemoglobin-synthesizing reticulocytes (Newman, 1982) but expression is lost in mature erythrocytes (Sutherland, 1981). Anti-CD71, Ber-79, was included in the Fifth International Workshop and Conference on Human Leucocyte Differentiation Antigens, and studies by a number of laboratories confirmed its reactivity with CD71 (Boeker, 1995).

Specificity

Antibody recognizes Human CD71 antigen.

Application

Detect CD71 using this Milli-Mark Anti-CD71-FITC Antibody, clone Ber-T9 validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human lymphocytes

Target description

190 kDa (Goding, 2002)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebekka Spellerberg et al.
Molecular therapy oncolytics, 27, 272-287 (2022-12-03)
Sodium iodide symporter (NIS) gene transfer for active accumulation of iodide in tumor cells is a powerful theranostic strategy facilitating both diagnostic and therapeutic application of radioiodide. In glioblastoma (GBM), the blood-brain barrier (BBB) presents an additional delivery barrier for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service