Skip to Content
Merck
All Photos(2)

Key Documents

MABC1765

Sigma-Aldrich

Anti-TfR1 (CD71) Antibody

purified antibody, clone 3F3-FMA, from mouse

Synonym(s):

Anti-CD71 antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€153.00
100 μL
€383.00

€153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€153.00
100 μL
€383.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€153.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F3-FMA, monoclonal

mol wt

calculated mol wt 84.87 kDa
observed mol wt ~90 kDa

species reactivity

human, mouse

packaging

antibody small pack of 100 μL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

Specificity

Clone 3F3-FMA is a mouse monoclonal antibody that detects human Transferrin receptor protein 1.

Immunogen

Purified membrane fractions from OCI-LY7 cells.

Application

Quality Control TestingEvaluated by Western Blotting in HT-1080 cell lysate.Western Blotting Analysis: A 1:500 dilution of this antibody detected TfR1 (CD71) in HT-1080 cell lysate.Tested ApplicationsFlow Cytometry Analysis: A representative lot detected TfR1 (CD71) in Flow Cytometry applications (Feng, H., et. al. (2020). Cell Rep. 30(10): 3411-3423).Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected TfR1 (CD71) in HT-1080 cells.Immunohistochemistry Applications: A representative lot detected TfR1 (CD71) in Immunohistochemistry applications (Feng, H., et. al. (2020). Cell Rep. 30(10): 3411-3423).Immunocytochemistry Analysis: A representative lot detected TfR1 (CD71) in Immunocytochemistry applications (Feng, H., et. al. (2020). Cell Rep. 30(10): 3411-3423). Western Blotting Analysis: A representative lot detected TfR1 (CD71) in Western Blotting applications (Feng, H., et. al. (2020). Cell Rep. 30(10): 3411-3423). Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TfR1 (CD71), clone 3F3-FMA, Cat. No. MABC1765, is a mouse monoclonal antibody that detects Tfr1 and is an excellent ferroptosis marker. It is tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

Transferrin receptor protein 1 (UniProt: Q62351; also known as TR, TfR, TfR1, Trfr, T9, p90, CD71) is encoded by the Tfrc (also known as Trfr) gene (Gene ID: 7037) in human. TfR1 is a single-pass type II membrane glycoprotein with a cytoplasmic domain (aa 1-67), a transmembrane domain (aa 68-88), and a long extracellular domain (aa 89-760). It can also be proteolytically cleaved at Arg100 to generate the soluble serum form (sTfR). TfR1 is involved in the uptake of iron via receptor mediated endocytosis into specialized endosomes where acidification leads to release of iron. TfR1 is reported to be essential for erythrocyte development and positive regulation of T and B cell proliferation through iron uptake. It also acts as a lipid sensor that regulates mitochondrial fusion by regulating the activation of the JNK pathway and degradation of mitofusin MFN2. Under conditions of low dietary levels of stearate (C18:0), it promotes the activation of JNK pathway that results in HUWE1-mediated ubiquitination and degradation of mitofusin MFN2 and inhibition of mitochondrial fusion. Clone 3F3-FMA is reported to be an effective ferroptosis-staining agent. It stains plasma membrane and the perinuclear region associated with the Golgi and the endosomal recycling compartment. (Ref.: Senilymaz, D., et al. (2015). Nature. 525(7567); 124-128; Feng, H., et al. (2020). Cell Rep. 30(10); 3411-3423).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service