Skip to Content
Merck
All Photos(1)

Key Documents

EZHFGF21

Millipore

Human FGF-21 ELISA Kit

measures FGF-21 in 50 μL serum, plasma or cell culture supernatent

Synonym(s):

ELISA Kit for FGF-21, FGF-21 ELISA, Human FGF-21 Detection Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
€984.00

€984.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
96 WELLS
€984.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

€984.00


Check Cart for Availability

Request a Bulk Order

Product Name

Human FGF-21 ELISA Kit, This Human FGF-21 ELISA Kit is used to measure & quantify FGF-21 levels in Metabolism & Inflammation & Immunology research.

Quality Level

species reactivity

human

packaging

kit of 1 × 96 wells

parameter

50 μL sample volume (4hr assay)

assay range

accuracy: 83-99%
standard curve range: 31.3-2000 pg/mL
inter-assay cv: <9%
intra-assay cv: <6%

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma (K2 EDTA)
sample type cell culture supernatant

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

General description

Recent studies have demonstrated that fibroblast growth factor 21 (FGF-21), a liver derived protein, is able to increase glucose uptake, as well as lower plasma triglyceride levels in an insulin-independent manner. FGF-21 suppresses glucagon release, thereby increasing insulin sensitivity, and reduces the amount of insulin production required by pancreatic β-cells. Unlike insulin, which works in a rapid, hormone-like manner, FGF-21 activity requires several hours, indicating changes in gene expression. While insulin functions through GLUT4 translocation, FGF-21 has been shown to act primarily through the upregulation of adipocyte GLUT1, a transporter that allows glucose to move from the blood into cells even in the absence of insulin. In addition, no significant weight gain was observed when FGF-21 was injected into animals, and mice that had been genetically engineered to overexpress FGF-21 weighed less and had less adipose tissue than the normal controls.

Specificity

No significant cross-reactivity to the cytokines and metabolic hormones tested was observed.

Application

Research Category
Metabolism
Research Sub Category
Diabetes - Type 2

Metabolic Syndrome
This Human FGF-21 ELISA Kit is used to measure & quantify FGF-21 levels in Metabolism & Inflammation & Immunology research.
This is a 3-hour assay requiring 50 μL of sample.
Used to detect/quantify: FGF-21

Other Notes

Please contact Technical Service for linearity of dilution.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Does the Human FGF-21 ELISA (EZHFGF21-19K) detect active or total FGF-21?

    1 answer
    1. We have not conducted specific testing to determine the assay's specificity to the active form of FGF-21. However, given that the antibodies used are polyclonal, it is likely that the assay detects Total FGF-21.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service