Skip to Content
Merck
All Photos(1)

Documents

EZHA

Millipore

Human Amylin ELISA Kit

measures and quantifies Amylin levels in 20 μL serum or plasma

Synonym(s):

Amylin ELISA, Human Amylin Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

product name

Human Amylin ELISA,

species reactivity

human

Quality Level

packaging

kit of 1 × 96 wells

parameter

20 μL sample volume (4hr assay)

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum

UniProt accession no.

application(s)

research use

assay range

inter-assay cv: 5.2-1.3
intra-assay cv: ±1.9 -0.7
standard curve range: 1-100 pM
accuracy: ±96 -2.5%

detection method

fluorometric (355nm/460nm)

shipped in

wet ice

storage temp.

2-8°C

Gene Information

General description

Amylin is a 37 amino acid hormone that is co-secreted with insulin from pancreatic beta cells in response to hyperglycemia.

Specificity

Used to detect/quantify: Amylin

Application

This Human Amylin ELISA is used to measure & quantify Amylin levels in Metabolism & Endocrine research.

50 μl plasma or cell culture supernatant 3.5 hour protocol, room temperature

Research Category: Metabolism

Research Sub-Category: Obesity, Metabolic Disorders

Packaging

96-well plate

Other Notes

This kit will be suitable for the measurement of amylin in rat and feline plasma, however, the precise percent of cross-reactivity is not determined.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service