Skip to Content
Merck
All Photos(2)

Documents

EZFITBACKF

Millipore

EZ-Fit® Manifold check valve

EZ-Fit® Manifold accessory, suitable for (General microbial analysis)

Synonym(s):

Membrane filter valve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

stainless steel

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 3 units

manufacturer/tradename

EZ-Fit®

input

sample type pharmaceutical(s)
sample type cosmetics
sample type beer
sample type bottled water

suitability

suitable for (General microbial analysis)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

Related Categories

General description

EZ-Fit® Manifold check valve is integrated into the flexible tubing connection to prevent the liquid back-flow from the vacuum system.

Application

The check valve prevents the liquid back-flow from the vacuum system in the EZ-Fit® Manifold. It enables efficient and contamination-free filtration

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service