Skip to Content
Merck
All Photos(1)

Key Documents

EZFITQUICKC

Millipore

EZ-Fit® Manifold quick connection

pkg of (1 plug and 1 connector), spare parts including one plug and connector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24
Pricing and availability is not currently available.

material

316L stainless steel

Sterilization Compatibility

autoclavable compatible

feature

spare parts including one plug and connector

manufacturer/tradename

EZ-Fit®

packaging

pkg of (1 plug and 1 connector)

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

General description

EZ-Fit® Manifold quick connection with one plug and one connector fits either end of the manifold pipe and helps to choose the direction of flow. The connector of the EZ-Fit® Manifold is connected to the vacuum system, and the plug is used to seal the other open end of the manifold pipe. The connector and plug create optimum vacuum and suction thereby, aiding in efficient filtration.

Application

The EZ-Fit® quick connection with one plug and one connector fits either end of the manifold pipe and helps to choose the direction of flow.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service