Skip to Content
Merck
All Photos(1)

Key Documents

EMS0008

Sigma-Aldrich

LysargiNase

suitable for peptide or protein digestion insolution or in gel

Synonym(s):

LysargiNase mirrors trypsin for protein C-terminal and methylation-site identification

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€219.00

€219.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
1 EA
€219.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.54

€219.00


Estimated to ship on21 May 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

form

solid

shipped in

ambient

Related Categories

General description

LysargiNase is a metalloproteinase found in the thermophilic archaea Methanosarcina acetivorans. It specifically cleaves before lysine and arginine residues in proteins. This cleavage results in peptides with molecular weights similar to tryptic peptides, but with N-terminal lysine or arginine residues that can be fragmented with b ion-dominated spectra. This can improve protein C-terminal peptide identification and several arginine rich phosphosite assignments. Unlike with trypsin, LysargiNase cleavage also occurs at methylated lysine and arginine, allowing detection of these epigenetic modifications.
LysargiNase is suitable for peptide or protein digestion in solution or in gel.

Application

LysargiNase is suitable for peptide or protein digestion in solution or in gel.
LysargiNase is suitable for peptide or protein digestion in solution or in gel. It is active under a variety of solvent conditions including 5 mM TCEP, 5% methanol, 5% acetonitrile, 0.8 M urea, 0.1% RapiGest, 1% deoxycholate, 0.2% SDS and 1% NP-40. It is active up to 55°C at pH 6 – 9. LysargiNase is reversibly inhibited by 1, 10-phenanthroline, EDTA and other calcium and zinc chelating agents. For peptide or protein digestion, a ratio of between 1:100 to 1:20 (w/w) enzyme:substrate is recommended.

Components

Each vial contains 20 µg LysargiNase, prepared by recombinant expression in E. coli and lyophilized from buffer.

Physical form

Each vial contains 20 µg LysargiNase, prepared by recombinant expression in E. coli and lyophilized from buffer. Upon reconstitution in 20 µL water, the enzyme is in 50 mM HEPES, 5 mM CaCl2, pH 7.5.

Storage and Stability

Store the lyophilized product at -20°C for up to 2 years. After reconstitution, the unused material can be stored in the freezer in aliquots. Avoid repeated freeze thaw cycles.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia Tallant et al.
The Journal of biological chemistry, 281(26), 17920-17928 (2006-04-22)
The metzincin clan encompasses several families of zinc-dependent metalloproteases with proven function both in physiology and pathology. They act either as broad spectrum protein degraders or as sheddases, operating through limited proteolysis. Among the structurally uncharacterized metzincin families are the
Le Zhang et al.
ACS chemical biology, 16(11), 2595-2603 (2021-11-05)
Methods for the selective labeling of biogenic functional groups on peptides are being developed and used in the workflow of both current and emerging proteomics technologies, such as single-molecule fluorosequencing. To achieve successful labeling with any one method requires that
Liana Tsiatsiani et al.
The FEBS journal, 282(14), 2612-2626 (2015-04-01)
Peptide-centered shotgun analysis of proteins has been the core technology in mass spectrometry based proteomics and has enabled numerous biological discoveries, such as the large-scale charting of protein-protein interaction networks, the quantitative analysis of protein post-translational modifications and even the
Pitter F Huesgen et al.
Nature methods, 12(1), 55-58 (2014-11-25)
To improve proteome coverage and protein C-terminal identification, we characterized the Methanosarcina acetivorans thermophilic proteinase LysargiNase, which cleaves before lysine and arginine up to 55 °C. Unlike trypsin, LysargiNase-generated peptides had N-terminal lysine or arginine residues and fragmented with b
Kazuya Tsumagari et al.
STAR protocols, 2(3), 100682-100682 (2021-08-12)
Characterization of protein termini is essential for understanding how the proteome is generated through biological processes such as post-translational proteolytic events. Here, we introduce a practical protocol for terminomics to achieve simple and sensitive N- and C-terminal peptide enrichment. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service