Skip to Content
Merck
All Photos(1)

Key Documents

DR1075

Sigma-Aldrich

Anti-TARDBP Mouse mAb (2E2-D3)

liquid, clone 2E2-D3, Calbiochem®

Synonym(s):

Anti-TAR DNA Binding Protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€442.00

€442.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€442.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€442.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E2-D3, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TARDBP(23435)

General description

Purified mouse monoclonal antibody. Recognizes the ~45 kDa TARDBP protein.
Recognizes the ~45 kDa TARDBP protein in A431 and HeLa cells and human leiomyosarcoma tissue.
This Anti-TARDBP Mouse mAb (2E2-D3) is validated for use in Immunoblotting, Immunocytochemistry, Paraffin Sections for the detection of TARDBP.

Immunogen

Full-length, recombinant human TARDBP (aa 1-261), expressed as a GST fusion protein.

Application




Immunoblotting (1-5 g/ml)
Immunocytochemistry (10 g/ml)
Paraffin Sections (5 &microg/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.7.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
293T cells
Positive Control
A431 cells, Human leiomyosarcoma tissue, HeLa cells

Other Notes

Ilieva, E,V., et al. 2010. Free Radic. Biol. Med.48, 1302.
Wils H, et al. 2010. Proc. Natl. Acad. Sci. U S A107 3858.
Foulds, P,G., et al. 2009. Acta Neuropathol.118 647.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service