Select a Size
About This Item
Skip To
form
liquid (Reagent)
Quality Level
packaging
vial of 500 μL
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
Northern blotting: suitable, Southern blotting: suitable
shipped in
dry ice
storage temp.
−20°C
1 of 4
This Item | |||
|---|---|---|---|
| form liquid (Reagent) | form liquid (Reagent) | form liquid (Reagent) | form - |
| packaging vial of 500 μL | packaging vial of 500 μL | packaging vial of 500 μL | packaging vial of 600 μL |
| manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore |
| growth mode N/A | growth mode N/A | growth mode N/A | growth mode N/A |
| technique(s) Northern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
Features and Benefits
Preparation Note
Other Notes
The DNA includes fragments ranging from 100-10,000 base pairs. The 1,000 and 3,000 base pair bands have increased intensity to serve as reference points.
Legal Information
Disclaimer
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| DPLUS-1K | 04065268021637 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


