Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
form
liquid (Reagent)
Quality Level
packaging
vial of 500 μL
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
Northern blotting: suitable, Southern blotting: suitable
shipped in
dry ice
storage temp.
−20°C
Features and Benefits
Preparation Note
Other Notes
The DNA includes fragments ranging from 100-3,000 base pairs. The 500 and 1,500 base pair bands have increased intensity to serve as reference points.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) Northern blotting: suitable, Southern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable |
| form liquid (Reagent) | form liquid (Reagent) | form liquid (Reagent) | form liquid (Reagent) |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| growth mode N/A | growth mode N/A | growth mode N/A | growth mode N/A |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| DPLUS-100H | 04065268021620 |



