Skip to Content
Merck
All Photos(1)

Documents

DEM711-10100

Millipore

Trial kit - NovaSeptum® GO; High Purity Bag Sampling System, 100ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104202

material

PureFlex polyethylene bag (Fluid contact layer, film)
TPE tubing (Thermoplastic elastomer)
polyester body
silicone septum (platinum-cured)
stainless steel body (ASTM® 316 L)

Quality Level

Agency

according to ISO ISO 11137
meets USP 88 Biological Reactivity

sterility

sterile; β-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

-20-50 °C temp. range (-4-122 °F) (1000 mL)
-80-50 °C temp. range (-112-122 °F) (50 mL to 250 mL (when used with a NovaSeptum® Case))
0.30 bar max. pressure (4.35 psi) (for 1000 mL)
0.50 bar max. pressure (7.25 psi) (up to 250 mL)

impurities

<2.15 bacterial endotoxins (LAL test, EU/device)

fitting

female Luer-Lok® outlet (3-piece)
male Luer-Lok® outlet (3-piece)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

room temp

General description

A range of NovaSeptum® GO small pack size kits available to trial a sterile sampling solution for your process. Choose from several collection container types: high purity bags, autoclavable bags, bottles or accurate volume syringes (5 containers per pack). The closed, sterile sampling method can significantly reduce risk of cross contamination. Ideal for sampling from aseptic and sterile processes, the NovaSeptum® GO sterile sampling system provides consistently representative samples, safely and securely. NovaSeptum® GO Accurate Volume Syringes are designed to take samples from 1 mL to 20 mL, enabling operators to directly dispense sample products with milliliter accuracy. Autoclavable bags are intended for applications where the unit is connected to a vessel that needs to be sterilized by autoclave. Bio-neutral plastic bottles or high purity bags are ideal for all tests and are available in a range of sizes. Use a trial kit in combination with a NovaSeptum® GO holder.

Application

Sterile sampling
Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications

Packaging

5x High Purity 100ml bag

Components

  • Septum: Platinum-cured silicone; Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Fluid contact layer (bag): Polyethylene film (PureFlex film)
  • Tubing: Thermoplastic elastomer (TPE)
  • Outlet Tubing: 3-piece Luer-Lok®, containing a male, female and an injection site

Preparation Note

All component materials, in contact with sampling liquid, meet the criteria for Class VI testing based on USP <88> Biological Reactivity, in vivo.
Units are integrity tested at regular intervals during manufacturing.
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.
Assembled under ISO Clean Room Class 8 conditions in a facility certified to ISO 14644-1 standards.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service