Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Check Cart for Availability | € 260,00 |
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
581, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
flow cytometry: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CD34(947)
Application
Inflammation & Immunology
Immunoglobulins & Immunology
Optimal working dilutions must be determined by the end user.
Biochem/physiol Actions
The antibody reacts with a single-chain 105-120 kDa heavily O-glycosylated transmembrane glycoprotein which is expressed on haematopoietic progenitor cells, vascular endothelium and some tissue fibroblasts. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. CD34 may also play a role in the adhesion of specific antigens to endothelium.
Physical form
Preparation Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source mouse | biological source mouse | biological source mouse | biological source mouse |
| species reactivity human | species reactivity human, nonhuman primates | species reactivity human, nonhuman primates | species reactivity human |
| conjugate unconjugated | conjugate phycoerythrin (R-PE) conjugate | conjugate unconjugated | conjugate Allophycocyanin conjugate |
| antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| clone 581, monoclonal | clone 581, monoclonal | clone 581, monoclonal | clone 4H11[APG], monoclonal |
| Gene Information human ... CD34(947) | Gene Information human ... CD34(947) | Gene Information human ... CD34(947) | Gene Information human ... CD34(947) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



![Monoclonal Anti-CD34-APC antibody produced in mouse clone 4H11[APG], purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/305/209/2c60cb51-14e3-48f2-ad1d-c8c23004cbb0/640/2c60cb51-14e3-48f2-ad1d-c8c23004cbb0.jpg)