Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700736

Sigma-Aldrich

Monoclonal Anti-CD34 antibody produced in mouse

clone 581, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Mucosialin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€224.00

€224.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€224.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€224.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

581, monoclonal

form

buffered aqueous solution

species reactivity

human, nonhuman primates

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD34(947)

General description

The mouse monoclonal antibody 581 reacts with CD34 (Mucosialin), a 110-115 kDa monomeric transmembrane phosphoglycoprotein expressed on hematopoietic progenitors cells and on the most pluripotential stem cells; it is gradually lost on progenitor cells. The antibody recognizes the class III CD34 epitope resistant to neuraminidase, chymopapain and glycoprotease.
HLDA V.; WS Code MA27.

Immunogen

Human mucosialin

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Wang et al.
Oncology letters, 11(5), 3091-3096 (2016-04-29)
The current study presents a case of cluster of differentiation (CD)56+ myeloid sarcoma in a patient that initially presented with skin lesions, and provides evidence for the clinical and differential diagnosis of myeloid sarcoma. The patient of the present case

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service