Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Check Cart for Availability | € 431,00 |
About This Item
€ 431,00
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
CHC5.9, monoclonal
species reactivity
bovine, amphibian, rat, pig, human
manufacturer/tradename
Chemicon®
technique(s)
immunohistochemistry: suitable, western blot: suitable
isotype
IgM
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CLTC(1213)
Immunogen
Application
Suitable for frozen sections
Western blotting KNOW SPECIES
Optimal working dilutions must be determined by the end user.
Epitope Tags & General Use
Organelle & Cell Markers
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Positive reactions using HeLa, SV-40 and RVF-SMC cultured cell lines.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| clone CHC5.9, monoclonal | clone 2E5, monoclonal | clone polyclonal | clone 4B12-1E3, monoclonal |
| Gene Information human ... CLTC(1213) | Gene Information human ... CLTC(1213) | Gene Information human ... CLTA(1211) | Gene Information human ... CLTB(1212) |
| species reactivity bovine, amphibian, rat, pig, human | species reactivity human | species reactivity rat, human | species reactivity human |
| biological source mouse | biological source mouse | biological source rabbit | biological source mouse |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form purified antibody | antibody form purified immunoglobulin | antibody form serum | antibody form purified immunoglobulin |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



