Skip to Content
Merck
All Photos(1)

Key Documents

AB9884

Sigma-Aldrich

Anti-Clathrin Light Chain Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€586.00

€586.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
€586.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€586.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CLTA(1211)

Specificity

Clathrin light chain a and b variants (CLC). By Western blot the antibody recognizes a doublet at ~35-40 kDa on rat brain extracts.

Application

This Anti-Clathrin Light Chain Antibody is validated for use in WB for the detection of Clathrin Light Chain.
Western blot: 1:50,000-1:100,000 on rat brain extracts.

Optimal working dilutions must be determined by end user.

Target description

~35-40 kDa

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Fan et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(32) (2021-08-08)
Pancreatic β cells operate with a high rate of membrane recycling for insulin secretion, yet endocytosis in these cells is not fully understood. We investigate this process in mature mouse β cells by genetically deleting dynamin GTPase, the membrane fission
Kazuki Obashi et al.
Nature communications, 14(1), 732-732 (2023-02-10)
Conformational changes in endocytic proteins are regulators of clathrin-mediated endocytosis. Three clathrin heavy chains associated with clathrin light chains (CLC) assemble into triskelia that link into a geometric lattice that curves to drive endocytosis. Structural changes in CLC have been
Yang Yang et al.
Developmental cell, 43(4), 507-521 (2017-11-22)
Assembly of the endocytic machinery is a constitutively active process that is important for the organization of the plasma membrane, signal transduction, and membrane trafficking. Existing research has focused on the stochastic nature of endocytosis. Here, we report the emergence
Mian Cao et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(49), 16544-16549 (2014-12-05)
Several proteins encoded by PD genes are implicated in synaptic vesicle traffic. Endophilin, a key factor in the endocytosis of synaptic vesicles, was shown to bind to, and be ubiquitinated by, the PD-linked E3 ubiquitin ligase Parkin. Here we report
Stephanie A Zlatic et al.
Molecular biology of the cell, 24(15), 2378-2388 (2013-06-14)
A role for clathrin in AP-3-dependent vesicle biogenesis has been inferred from biochemical interactions and colocalization between this adaptor and clathrin. The functionality of these molecular associations, however, is controversial. We comprehensively explore the role of clathrin in AP-3-dependent vesicle

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service