Skip to Content
Merck

Skip To

CB1052

Anti-TUBB2A Mouse mAb (2B2)

liquid, clone 2B2, Calbiochem®

Synonym(s):

Anti-Tubulin β2A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 381,00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
2B2, monoclonal
Species reactivity:
human, mouse, rat
Application:
Citations:
-

€ 381,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B2, monoclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoblotting (1-5 µg/mL)
Paraffin Sections (1.5 µg/mL))

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB2A(7280)

General description

Purified mouse monoclonal antibody. Recognizes the ~48-50 kDa TUBB2A protein.
Recognizes the ~48-50 kDa TUBB2A protein in PC-12, Raw 264.7, Jurkat, and NIH3T3 cells and human spleen tissue.
This Anti-TUBB2A Mouse mAb (2B2) is validated for use in Immunoblotting, Paraffin Sections for the detection of TUBB2A.

Immunogen

Full-length, recombinant, human TUBB2A (aa 1-446), expressed as a GST fusion protein

Application





Immunoblotting (1-5 g/ml)
Paraffin Sections (1.5 g/ml)

Physical form

In PBS, pH 7.2.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
293T cells
Positive Control
PC-12 cells, Raw 264.7 cells, Jurkat cells, NIH3T3 cells, Human spleen tissue

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DR1075ST1705OP145
Gene Information

human ... TUBB2A(7280)

Gene Information

human ... TARDBP(23435)

Gene Information

human ... TOMM20(9804)

Gene Information

human ... MDM2(4193)

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

species reactivity

human, mouse, rat

species reactivity

human

species reactivity

human, rat, mouse

species reactivity

human

clone

2B2, monoclonal

clone

2E2-D3, monoclonal

clone

4F3, monoclonal

clone

4B2C1.11, monoclonal

isotype

IgG2a

isotype

IgG1

isotype

IgG1

isotype

IgG1


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters