BE-10X2ML-B
Anti-Jkb (clone MS-8) Blood Grouping Reagent
BIOSCOT™ Anti-Jkb from the clone MS-8, Human IgM, Blood Grouping Reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116102
Recommended Products
biological source
human
Quality Level
clone
MS-8, monoclonal
packaging
pkg of 10 × 2 mL
manufacturer/tradename
BIOSCOT™
storage condition
do not freeze
concentration
30-56 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
7.1-7.3
isotype
IgM
application(s)
clinical testing
shipped in
ambient
storage temp.
2-8°C
General description
Contains 0.1% (w/v) Sodium Azide;Filtered through a 0.22μm terminal filter;Shelf Life: 36 months from date of filtration;Product formulation meets the requirements of Annex IV of the EU IVD directive to support CE registration;For In Vitro use only .
Application
Tube technique
Vialled, Labeled
Legal Information
BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service