Skip to Content
Merck
All Photos(1)

Key Documents

AX0442

Supelco

Reagent Alcohol

82.0-86.0% ethanol basis, Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07
MDL number:
UNSPSC Code:
12352104
grade:
reagent
Pricing and availability is not currently available.

Product Name

Alcohol, Reagent

grade

reagent

Quality Level

Assay

82.0-86.0% ethanol basis

form

liquid

composition

isopropyl alcohol, 4.0-6.0%
methanol, 3.5-5.5%
Water, 7-8%

color

APHA: ≤20

transition temp

flash point 17 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent

Physical form

To each 100 parts of SD 3A Alcohol are added 5 parts of Isopropyl Alcohol

Analysis Note

Assay (Ethanol): 82.0-86.0%
Assay (Isopropyl alcohol): 4.0-6.0%
Assay (Methanol): 3.5-5.5%
Color (APHA): 20 max
Form: Clear liquid
Water (H2O): 7-8%

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service