Skip to Content
Merck
All Photos(1)

Key Documents

AP183B

Sigma-Aldrich

Goat Anti-Rat IgG Antibody, biotin-SP conjugate, Species Adsorbed

1.5 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
€363.00

€363.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 μL
€363.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€363.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

concentration

1.5 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Detect Rat IgG using this Goat anti-Rat IgG Antibody, biotin-SP conjugate, Species Adsorbed validated for use in ELISA & WB.
EIA and Western blots: 120,000-1:400,000.

Enzyme Immunohistochemistry: 1:500-1:5,000.

Flow cytometry and Fluorescence immunohistochemistry: 1:200-1:1000

Optimal working dilutions must be determined by end user.

Physical form

Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15 mg/mL BSA, and 0.05% sodium azide.

RECONSTITUTION:

Reconstitute with 500 μL of sterile distilled water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Hyer et al.
The European journal of neuroscience, 43(5), 703-709 (2016-01-12)
Our understanding of postpartum anxiety (PPA) in fathers is limited, despite the negative consequences of anxiety on the father and child. Offspring contact reduces PPA in mothers; however, parallel investigations in fathers has gone unaddressed. Adult neurogenesis in the dentate
Antagonism of the histamine H4 receptor reduces LPS-induced TNF production in vivo.
Cowden, JM; Yu, F; Challapalli, M; Huang, JF; Kim, S; Fung-Leung, WP; Ma, JY; Riley et al.
Inflammation Research null
Christine R Collins et al.
eLife, 9 (2020-12-09)
Red blood cell (RBC) invasion by malaria merozoites involves formation of a parasitophorous vacuole into which the parasite moves. The vacuole membrane seals and pinches off behind the parasite through an unknown mechanism, enclosing the parasite within the RBC. During
Abhinay Ramaprasad et al.
eLife, 11 (2022-12-29)
The malaria parasite Plasmodium falciparum synthesizes significant amounts of phospholipids to meet the demands of replication within red blood cells. De novo phosphatidylcholine (PC) biosynthesis via the Kennedy pathway is essential, requiring choline that is primarily sourced from host serum
Xiaohong Xiang et al.
Frontiers in medicine, 8, 746346-746346 (2021-11-09)
Kidney injury associated with cold storage/transplantation is a primary factor for delayed graft function and poor outcome of renal transplants. p53 contributes to both ischemic and nephrotoxic kidney injury, but its involvement in kidney cold storage/transplantation is unclear. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service