Skip to Content
Merck
All Photos(1)

Key Documents

MABF2057

Sigma-Aldrich

Anti-minicore HCV Antibody, clone Neo4

clone Neo4, from mouse

Synonym(s):

HCV, Hepatitis C virus minocores

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€131.00
100 μG
€343.00

€131.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€131.00
100 μG
€343.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€131.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Neo4, monoclonal

species reactivity

virus

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

General description

Hepatitis C virus (HCV) is a small (~55 65 nm) in size), enveloped, positive-sense single-stranded RNA virus that is a causative factor for hepatitis C and hepatocellular carcinoma. HCV expresses several proteins that promote viral replication. Genome polyprotein of HCV is a core protein that packages viral RNA to form a viral nucleocapsid, and promotes virion budding. It also modulates viral translation initiation by interacting with HCV IRES and 40S ribosomal subunit. It prevents the establishment of cellular antiviral state by blocking the interferon-alpha/beta (IFN-alpha/beta) and IFN-gamma signaling pathways and by inducing human STAT1 degradation. Envelope (E1 and E2) glycoproteins of HCV form a heterodimer, which is involved in virus attachment to the host cell, virion internalization through clathrin-dependent endocytosis and fusion with host membrane. P7 protein, a heptameric ion channel protein, is essential for infectivity. HCV also contains NS2-3, which acts as a cysteine protease responsible for the autocatalytic cleavage of NS2-NS3. It undergoes self-inactivation following maturation. NS3 protein of HCV displays serine protease, NTPase, and RNA helicase activities. NS4B protein is shown to induce a specific membrane alteration that serves as a scaffold for the virus replication complex. NS5A protein is a component of the replication complex and is involved in RNA-binding. It is reported to mediate interferon resistance by interacting with and inhibiting human EIF2AK2/PKR. HCV also expresses a family of small proteins that are derived from the core gene and are known as minocores. These minicores lack the N-terminus, but contain the C-terminal portion of the mature p21 core protein. They are associated with higher risk of hepatocellular carcinoma, insulin resistance, and failure of interferon-based therapy.

Specificity

Clone Neo4 detects HCV core and minicore proteins in cells infected with HCV virus.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal region of Hepatitis C virus (HCV) core protein.

Application

Anti-minicore HCV, clone Neo4, Cat. No. MABF2057, is a mouse monoclonal antibody that detects intracellular and secreted HCV core and minicore proteins in HCV infected cells and has been tested for use in Immunofluorescence and Western Blotting.
Research Category
Inflammation & Immunology
Western Blotting Analysis: A representative lot detected minicore HCV in Western Blotting applications (Eng, F.J., et. al. (2018). Hepatol Commun. 2(1):21-28).

Immunofluorescence Analysis: A representative lot detected minicore HCV in Immunofluorescence applications (Eng, F.J., et. al. (2018). Hepatol Commun. 2(1):21-28).

Quality

Evaluated by Western Blotting in extracts of HEK293T cells transfected with plasmids expressing only HCV p21 core and 91 minicore proteins.

Western Blotting Analysis: 4 ug/mL of this antibody detected minicore HCV in extracts of HEK293T cells transfected with plasmids expressing only HCV p21 core and 91 minicore proteins.

Target description

~ 8 and 21 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service