Skip to Content
Merck
All Photos(1)

Key Documents

AP180B

Sigma-Aldrich

Donkey Anti-Goat IgG Antibody, biotin-SP conjugate, Species Adsorbed

1 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
€376.00

€376.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
500 μL
€376.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€376.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

donkey

Quality Level

conjugate

biotin conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

goat

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Specific for Goat IgG.

Application

Donkey anti-Goat IgG Antibody, biotin-SP conjugate, Species Adsorbed detects level of Donkey Goat IgG & has been published & validated for use in ELISA, FC, IF, IH & WB.
EIA: 1:20,000-1:400,000
Western blots using enzyme-conjugated streptavidin: 1:20,000-1:400,000.
Immunohistochemistry: 1:500-1:5,000
Flow cytometry: 1:200-1:1,000
Fluorescence Immunohisto/cytochemistry: 1:200-1:1,1000
Optimal working dilutions must be determined by end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Physical form

Immunoaffinity Purified and Immunodepletion
Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 15 mg/mL BSA, and 0.05% sodium azide.

RECONSTITUTION:
Reconstitute with 500 μL of sterile distilled water.

Storage and Stability

Maintain lyophilized product at 2–8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2–8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Y Yu et al.
The Journal of international medical research, 38(2), 583-592 (2010-06-03)
The effect of 2,2'-dipyridyl (DP) on cerebral vasospasm was investigated in a double-injection rabbit model of subarachnoid haemorrhage (SAH). Thirty-six animals were divided between four groups: control (sham-operated), SAH (model alone), SAH + DP (the SAH model in which DP
Bastian Henkel et al.
Chemical senses, 42(1), 25-35 (2016-09-23)
In most mammals, the vomeronasal system detects a variety of (semio)chemicals that mediate olfactory-driven social and sexual behaviors. Vomeronasal chemosensation depends on G protein-coupled receptors (V1R, V2R, and FPR-rs) that operate at remarkably low stimulus concentrations, thus, indicating a highly
Yulia Shamis et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(16), E2020-E2029 (2015-04-08)
Zfp57 is a maternal-zygotic effect gene that maintains genomic imprinting. Here we report that Zfp57 mutants exhibited a variety of cardiac defects including atrial septal defect (ASD), ventricular septal defect (VSD), thin myocardium, and reduced trabeculation. Zfp57 maternal-zygotic mutant embryos
Alice E Adriaenssens et al.
Cell metabolism, 30(5), 987-996 (2019-08-27)
Ambiguity regarding the role of glucose-dependent insulinotropic polypeptide (GIP) in obesity arises from conflicting reports asserting that both GIP receptor (GIPR) agonism and antagonism are effective strategies for inhibiting weight gain. To enable identification and manipulation of Gipr-expressing (Gipr) cells
Long-term exercise is a potent trigger for ?FosB induction in the hippocampus along the dorso-ventral axis.
Nishijima, T; Kawakami, M; Kita, I
Testing null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service