Skip to Content
Merck

Skip To

ABS986

Anti-PCTP Antibody

serum, from rabbit

Synonym(s):

Phosphatidylcholine transfer protein, PC-TP, START domain-containing protein 2, StARD2, StAR-related lipid transfer protein 2

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
2


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PCTP(58488)

General description

Phosphatidylcholine transfer protein (PCTP), also known as PC-TP, START domain-containing protein 2, StARD2, StAR-related lipid transfer protein 2, and encoded by the gene symbol PCTP/ STARD2, belongs to the steroidogenic acute regulatory protein related transfer (START) domain superfamily of hydrophobic ligand-binding proteins. PCTP has been shown to play an important role in the transfer of phosphatidylcholine between membranes. The crystal structures of human PC-TP in complex with dilinoleoyl-PtdCho or palmitoyl-linoleoyl-PtdCho suggests that a single well-ordered PtdCho molecule occupies a centrally located tunnel. These structures have provided a basis for rationalizing the specificity of PC-TP for PtdCho and may identify common features used by START proteins to bind their hydrophobic ligands. Additionally, PCTP has been demonstrated to interact with ACOT13/THEM2. PCTP is located in the cytoplasm with highest expression in liver, placenta, testis, kidney and heart. Low levels of PCTP are expressed in brain and lung.
~33 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human PCTP.

Application

Anti-PCTP is an antibody against PCTP for use in WB, ICC, IP.
Western Blotting Analysis: A representative lot detected PCTP in cytosolic fractions isolated from the liver of a control subject, a patient with intrahepatic brown pigment stones, and a patient with obstructive cholestasis (Shoda, J., et al. (2001). Hepatology. 33(5):1194-1205).
Western Blotting Analysis: A representative lot detected PCTP in siRNA-transfected, serum-starved HEK 293E cells, Pctp+/+ liver lysates, Tsc2−/− MEFs, Liver homogenates from chow diet–fed Pctp+/+ (n = 4) and Pctp−/− (n = 4) mice (Ersoy, B.A., et al. (2013). Science Signaling. 286(6):1-11).
Immunocytochemistry Analysis: A representative lot detected PCTP in HEK293T cells (Kanno, K., et al. (2007). J. Biol. Chem. 282:30728-30736).
Immunoprecipitation Analysis: A representative lot immunoprecipitated PCTP in Pctp+/+ liver lysates (Ersoy, B.A., et al. (2013). Science Signaling. 286(6):1-11).

Analysis Note

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PCTP in 5 µg of HepG2 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA022979MABS1220NE1040
biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

antibody form

serum

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

Gene Information

human ... PCTP(58488)

Gene Information

human ... PCTP(58488)

Gene Information

human ... CETP(1071)

Gene Information

human ... PLTP(5360)

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters