Skip to Content
Merck
All Photos(3)

Key Documents

HPA022979

Sigma-Aldrich

Anti-PCTP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-STARD2, Anti-phosphatidylcholine transfer protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LAAGSFSEEQFWEACAELQQPALAGADWQLLVETSGISIYRLLDKKTGLYEYKVFGVLEDCSPTLLAD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCTP(58488)

General description

The gene PCTP (phosphatidylcholine transfer protein) is mapped to human chromosome 17q21-22. It belongs to the steroidogenic acute regulatory protein related transfer (START) domain superfamily. The protein is present in the cytoplasm.

Immunogen

phosphatidylcholine transfer protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PCTP (phosphatidylcholine transfer protein) is needed for the intermembrane exchange of phosphatidylcholines. Gene variants of PCTP affect the LDL (low density lipoprotein) peak particle size.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75640

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Cohen et al.
Biochimica et biophysica acta, 1447(2-3), 265-270 (1999-11-05)
Phosphatidylcholine transfer protein (PC-TP) is a cytosolic protein that catalyzes intermembrane transfer of phosphatidylcholines in vitro. We have cloned a cDNA encoding the human ortholog of PC-TP and have determined its tissue-specific expression as well as genomic organization. Radiation hybrid
Steven L Roderick et al.
Nature structural biology, 9(7), 507-511 (2002-06-11)
Phosphatidylcholines (PtdChos) comprise the most common phospholipid class in eukaryotic cells. In mammalian cells, these insoluble molecules are transferred between membranes by a highly specific phosphatidylcholine transfer protein (PC-TP) belonging to the steroidogenic acute regulatory protein related transfer (START) domain
G Dolley et al.
Atherosclerosis, 195(2), 297-302 (2007-02-03)
The small, dense LDL phenotype is associated with an increased cardiovascular disease risk. A genome-wide scan performed on 236 nuclear families of the Quebec Family Study (QFS) revealed a quantitative trait locus (QTL) affecting LDL peak particle size (LDL-PPD) and
A potential role for lysophosphatidylcholine in the delivery of long chain polyunsaturated fatty acids to the fetal circulation.
Véronique Ferchaud-Roucher et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1864(3), 394-402 (2018-12-21)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service