Skip to Content
Merck
All Photos(2)

Key Documents

ABN428

Sigma-Aldrich

Anti-Fezf2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Fez family zinc finger protein 2, Forebrain embryonic zinc finger-like protein 2, Zinc finger protein 312, Zinc finger protein Fez-like

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FEZF2(55079)

General description

Fezf2 is a transcriptional repressor protein. It is required for the specification of corticospinal motor neurons and other subcerebral projection neurons. FeZf2 also plays a role in the layer and subcortical specific patterning of the subcortical projections and axonal fasciculations that develop in the brain. Fezf2 also controls the development of the dendritic arborization and spines of the large layer V pyramidal neurons. Thus biological processes involving Fezf2 include neuronal patterning, differentiation, neurogenesis, axon guidance, fasciculation, cortex development, anterior/posterior patterning, dendrite development, and locomotory behavior, and it was recently identified as a multipotent progenitor marker for neocortical projection neurons, astrocytes, and oligodendrocytes.

Immunogen

Epitope: Engrailed homology 1 repressor domain
KLH-conjugated linear peptide corresponding to the Engrailed homology 1 repressor domain of human Fezf2.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Fezf2 in rat spinal cord tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Fezf2 Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of Fezf2.

Quality

Evaluated by Western Blotting in mouse E13-14 brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Fezf2 in 10 µg of mouse E13-14 brain tissue lysate.

Target description

~50 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erna A van Niekerk et al.
Cell reports methods, 2(7), 100255-100255 (2022-07-27)
Neuronal cultures provide a basis for reductionist insights that rely on molecular and pharmacological manipulation. However, the inability to culture mature adult CNS neurons limits our understanding of adult neuronal physiology. Here, we report methods for culturing adult central nervous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service