Skip to Content
Merck
All Photos(1)

Key Documents

900774

Sigma-Aldrich

Di-PDI

Synonym(s):

2,2′,9,9′-Tetrakis(1-pentylhexyl)-[5,5′-bianthra[2,1,9-def:6,5,10-de f′]diisoquinoline]-1,1′,3,3′,8,8′,10,10′(2H,2′H,9H,9′H)-octone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€747.00

€747.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€747.00

About This Item

Empirical Formula (Hill Notation):
C92H106N4O8
CAS Number:
Molecular Weight:
1395.85
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

€747.00


Check Cart for Availability

Request a Bulk Order

description

Band gap: 2.06 eV

Quality Level

Assay

≥97%

form

powder

solubility

chloroform: soluble
dichlorobenzene: soluble

Orbital energy

HOMO -5.81 eV 
LUMO -3.75 eV 

SMILES string

N1(C(=O)c2c3c4c(c%12c%13c(c4ccc3C1=O)ccc%14c%13c(cc%12)C(=O)N(C%14=O)C(CCCCC)CCCCC)c(c2)c5c6c7c(c9c%10c6ccc%11c%10c(cc9)C(=O)N(C%11=O)C(CCCCC)CCCCC)ccc8c7c(c5)C(=O)N(C8=O)C(CCCCC)CCCCC)C(CCCCC)CCCCC

InChI key

SROWQESEJZYBTR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Zang et al.
Advanced materials (Deerfield Beach, Fla.), 26(32), 5708-5714 (2014-06-20)
High-performance non-fullerene OSCs with PCEs of up to ca. 6.0% are demonstrated based on PBDTT-F-TT polymer and a molecular di-PBI acceptor through comprehensive molecular, interfacial, and device engineering. Impressive PCEs can also be retained in devices with relatively thick BHJ
Wei Jiang et al.
Chemical communications (Cambridge, England), 50(8), 1024-1026 (2013-12-07)
A series of bay-linked perylene bisimides as non-fullerene acceptors for organic solar cells are designed. The best power conversion efficiency up to 3.63% based on s-diPBI (1b) is demonstrated by fine-tuning optoelectronic properties resulting from different degrees of twisting and
A perylene bisimide derivative with a LUMO level of -4.56 eV for non-fullerene solar cells.
Yu Y, et al.
Journal of Material Chemistry C, 4(19), 4134-4137 (2016)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service