Skip to Content
Merck

Skip To

ABE91

Anti-JMJD8 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

JMJD, JmjC domain-containing protein 8, Jumonji domain-containing protein 8

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 529,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

€ 529,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, pig, horse

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), canine (based on 100% sequence homology), equine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... JMJD8(339123)

General description

Covalent modification of histones plays critical role in regulating chromatin structure and transcription. JMJD8 is one member of the family of JmjC domain-containing histone demethylation (JHDM) enzymes. These proteins remove specific methyl marks from histones. Histone demethylation by JHDM proteins requires cofactors Fe(II) and alpha-ketoglutarate. Contributions of histone demethylase activity to tumor development, decreases in cell proliferation, and hormone-dependent transcriptional activation have been observed. While two isoforms of JMJD8 have been described, further study is still required to clarify the roles and mechanisms of JMJD8 among other JHDM protein family members.
~37 kDa observed. Two isoforms with molecular weights of ~34 kDa and ~37 kDa have been described. An uncharacterized band at ~160 kDa may be observed in some lysates.

Immunogen

KLH-conjugated linear peptide corresponding to human JMJD8.

Application

Anti-JMJD8 Antibody is a Rabbit Polyclonal Antibody for detection of JMJD8 also known as Jumonji domain-containing protein 8 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract
Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 1 mg/mL of this antibody dected JMJD8 in 10 mg of HeLa nuclear extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1300005MABE222ABE498
description

from rabbit, purified by affinity chromatography

description

IgG fraction of antiserum, buffered aqueous solution

description

clone 8B7.1, from rabbit

description

from rabbit, purified by affinity chromatography

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

technique(s)

western blot: suitable

technique(s)

indirect ELISA: 1:1000, western blot: 1:100-1:500

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

western blot: suitable

species reactivity

human, pig, horse

species reactivity

human

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters