Skip to Content
Merck
All Photos(1)

Key Documents

ABE498

Sigma-Aldrich

Anti-Lysine-specific demethylase 7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Lysine-specific demethylase 7, JmjC domain-containing histone demethylation protein 1D

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€279.00

€279.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€279.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€279.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee, giant panda, mouse, canine, rhesus macaque, marmoset

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... JHDM1D(80853)

General description

Lysine-specific demethylase 7, also known as JmjC domain-containing histone demethylation protein 1D, and encoded by the gene JHDM1D/KDM7/KIAA1718, is an important histone demethylase particularly for brain development. JHDM1D specifically demethylates dimethylated ′Lys-9′ and ′Lys-27′ (H3K9me2 and H3K27me2, respectively) of histone H3 and monomethylated histone H4 ′Lys-20′ residue (H4K20Me1), thereby playing a central role in histone code. JHDM1D also specifically binds trimethylated ′Lys-4′ of histone H3 (H3K4me3), affecting histone demethylase specificity: in presence of H3K4me3, it has no demethylase activity toward H3K9me2, while it has high activity toward H3K27me2. JHDM1D also demethylates H3K9me2 in absence of H3K4me3 and JHDM1D has activity toward H4K20Me1 only when the nucleosome is used as a substrate and when not the histone octamer is used as substrate. JHDM1D is located in the nucleus and widely expressed but lowered expression is observed in glial cells and lymphoid cells. JHDM1D demonstrates tumor suppression by regulating angiogenesis factors when overexpression under starvation conditions in certain cancers, thus JHDM1D is being examined as a potential anti-cancer target.

Specificity

This antibody is expected to cross react with two known isoforms of Lysine-specific demethylase 7 based upon immunogen design.

Immunogen

KLH-conjugated linear peptide corresponding to human Lysine-specific demethylase 7.

Application

Detect Lysine-specific demethylase 7 using this rabbit polyclonal antibody, Anti-Lysine-specific demethylase 7 Antibody validated for use in western blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Lysine-specific demethylase 7 in 10 µg of HEK293 cell lysate.

Target description

~110 kDa observed. Uniprot describes two isoforms at 107 kDa (isoform 1) and 92 kDa (isoform 2).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service