Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
200 μL | Check Cart for Availability | € 402,00 |
About This Item
biological source
rabbit
Quality Segment
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, pig
species reactivity (predicted by homology)
mouse (based on 100% sequence homology), Xenopus (based on 100% sequence homology), porcine (based on 100% sequence homology), chicken (based on 100% sequence homology), zebrafish (based on 100% sequence homology), rat (based on 100% sequence homology)
technique(s)
cell based assay: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
phosphorylation (pTyr1161/pTyr1165/pTyr1166)
Gene Information
human ... IGF1R(3480)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Signaling
Insulin/Energy Signaling
Insulin/Energy Signaling
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
IGF1 untreated and treated HEK293 cell lysate
Western Blot Analysis: A 1:200 dilution of this antibody detected IGF-1R in IGF1 treated HEK293 cell lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source rabbit | biological source rabbit | biological source rabbit | biological source mouse |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified antibody |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| Gene Information human ... IGF1R(3480) | Gene Information human ... IGF1R(3480) | Gene Information human ... IGF1R(3480) | Gene Information human ... IGF1R(3480) |
| clone polyclonal | clone polyclonal | clone polyclonal | clone αIR3, monoclonal |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



