Skip to Content
Merck
All Photos(2)

Key Documents

ABC837

Sigma-Aldrich

Anti-ARC Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nucleolar protein 3, Apoptosis repressor with CARD, Muscle-enriched cytoplasmic protein, Myp, Nucleolar protein of 30 kDa, Nop30, ARC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€335.00

€335.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€335.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€335.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOL3(8996)

General description

Nucleolar protein 3 (NOL3), also known as Apoptosis repressor with CARD or Muscle-enriched cytoplasmic protein (Myp) or Nucleolar protein of 30 kDa (Nop30) and encoded by the gene name NOL3 or ARC contains one CARD domain. Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD) containing molecules and a caspase family of proteases. CARD domain containing cell death regulators include RAIDD, Apaf-1, caspase-9, and caspase-2. A novel CARD domain containing protein was recently identified and designated ARC for apoptosis repressor with CARD. ARC interacts with caspase-2 and –8 and inhibits enzymatic activity of caspase-8. ARC suppresses apoptosis induced by cell death adapters FADD and TRADD and by cell death receptors Fas, TNFR-1 and DR3. The messenger RNA of ARC is primarily expressed in skeletal muscle and cardiac tissue.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to amino acids 191-208 at the C-terminus of human ARC.

Application

Anti-ARC Antibody detects level of ARC & has been published & validated for use in ARC.
Immunohistochemistry Analysis: 5 μg/mL from a representative lot detected ARC in human heart and skeletal muscle tissues.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in HeLa, KB, and A549 cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ARC in HeLa, KB, and A549 cell lysates.

Target description

~24 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian K Chung et al.
Journal of autoimmunity, 77, 45-54 (2016-10-30)
Primary biliary cholangitis (PBC) and primary sclerosing cholangitis (PSC) are immune-mediated biliary diseases that demonstrate prominent and restricted genetic association with human leukocyte antigen (HLA) alleles. In PBC, anti-mitochondrial antibodies (AMA) are specific and used as diagnostic biomarkers. PSC-relevant auto-antibodies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service